missing translation for 'onlineSavingsMsg'
Learn More

Methyl 6-bromohexanoate, 96+%

Product Code. 10687664 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 10687664

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14273-90-6 | C7H13BrO2 | 209.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14273-90-6
Molecular Formula C7H13BrO2
Molecular Weight (g/mol) 209.08
MDL Number MFCD00187528
InChI Key KYLVAMSNNZMHSX-UHFFFAOYSA-N
Synonym 6-bromohexanoic acid methyl ester, hexanoic acid, 6-bromo-, methyl ester, methyl-6-bromohexanoate, methyl6-bromohexanoate, pubchem16910, methyl 6-bromo-hexanoate, 6-bromo-hexanoic acid methyl ester, 6-bromo-hexanoic acid-methyl ester, 6-bromo-henxanoic acid methyl ester, hexanoic acid,6-bromo-, methyl ester
PubChem CID 4170292
IUPAC Name methyl 6-bromohexanoate
SMILES COC(=O)CCCCCBr

Specifications

Color Colorless to Yellow
Density 1.3160g/mL
Boiling Point 150.0°C (50.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula Br(CH2)5C(O)OCH3
Refractive Index 1.4630 to 1.4650
Quantity 5 g
Specific Gravity 1.316
Formula Weight 209.08
Percent Purity 96+%
Physical Form Liquid
Chemical Name or Material Methyl 6-bromohexanoate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.