missing translation for 'onlineSavingsMsg'
Learn More

Methyl 6-chloropicolinate, 98%, BLD Pharm™

Product Code. 31424397
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
100 g
500 g
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31424397 10 g
31415129 500 g
31414699 5 g
31423326 25 g
31421506 1 g
31417464 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 31424397 Supplier BLD Pharm Supplier No. BD10875310g
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 6636-55-1
Molecular Formula C7H6ClNO2
MDL Number MFCD00155391
Quantity 10 g
SMILES O=C(C1=NC(Cl)=CC=C1)OC
Molecular Weight (g/mol) 171.58
Percent Purity 98%
Chemical Name or Material Methyl 6-chloropicolinate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.