missing translation for 'onlineSavingsMsg'
Learn More

Methyl 6-formylpyridine-2-carboxylate, Thermo Scientific™

Product Code. 10476964
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10476964 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10476964 Supplier Thermo Scientific Maybridge Supplier No. MO07188DA
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 69950-65-8
Packaging Amber Glass Bottle
Molecular Formula C8H7NO3
Quantity 1 g
Synonym methyl 6-formyl-2-pyridinecarboxylate, methyl 6-formylpicolinate, 6-formyl-2-pyridine carboxylic acid methyl ester, 2-pyridinecarboxylic acid, 6-formyl-, methyl ester, methyl6-formyl-2-pyridinecarboxylate, 2-formyl-6-methoxycarbonyl pyridine, 6-formylpyridine-2-carboxylic acid methyl ester, acmc-1b7li, 6-formyl-pyridine-2-carboxylic acid methyl ester
InChI Key CERBENZCBVYKEF-UHFFFAOYSA-N
SMILES COC(=O)C1=CC=CC(=N1)C=O
IUPAC Name methyl 6-formylpyridine-2-carboxylate
Molecular Weight (g/mol) 165.148
PubChem CID 12575850
Formula Weight 165.15
Percent Purity 97%
Chemical Name or Material Methyl 6-formylpyridine-2-carboxylate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.