missing translation for 'onlineSavingsMsg'
Learn More

Methyl 7-phenylheptanoate, 98%

Product Code. p-7047184
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11314825

Brand: Thermo Scientific Alfa Aesar L17905.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 101689-18-3 | C14H20O2 | 220.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 101689-18-3
Molecular Formula C14H20O2
Molecular Weight (g/mol) 220.31
MDL Number MFCD02093971
InChI Key FZSNATXOGRAOIL-UHFFFAOYSA-N
Synonym acmc-20akft, benzeneheptanoic acid,methyl ester, 7-phenylheptanoic acid methyl ester
PubChem CID 7010318
IUPAC Name methyl 7-phenylheptanoate
SMILES COC(=O)CCCCCCC1=CC=CC=C1

Specifications

Boiling Point ∼155°C (1.5mmHg)
Refractive Index 1.4946
Quantity 1 g
Beilstein 2694776
Formula Weight 220.31
Percent Purity 98%
Chemical Name or Material Methyl 7-phenylheptanoate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.