missing translation for 'onlineSavingsMsg'
Learn More

Methyl acetate, CHROMASOLV™, for HPLC, ≥99.8%, Honeywell Riedel-de Haën™

Product Code. 15615620
Change view
Click to view available options
Quantity:
250 mL
2.5 L
Unit Size:
2.5 L
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15615620 250 mL 250mL
15605620 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15615620 Supplier Honeywell-Chromasolv Supplier No. 45999250ML
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CHROMASOLV™, for HPLC,≥99.8%

  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-20-9
Molecular Formula C3H6O2
Molecular Weight (g/mol) 74.079
MDL Number MFCD00008711
InChI Key KXKVLQRXCPHEJC-UHFFFAOYSA-N
Synonym tereton, devoton, acetic acid, methyl ester, methyl ethanoate, acetic acid methyl ester, methylacetat, acetate de methyle, methylacetaat, methyl acetic ester, octan metylu
PubChem CID 6584
ChEBI CHEBI:77700
IUPAC Name methyl acetate
SMILES CC(=O)OC

Specifications

Density 0.934g/mL at 25°C
Flash Point −13°C (8.6°F)
Packaging Glass bottle
Linear Formula CH3COOCH3
Quantity 250 mL
UN Number UN1231
Beilstein 1736662
Vapor Density 2.55 (vs air)
Formula Weight 74.08g/mol
Percent Purity ≥99.8% (GC)∼∼≥99.8%
Chemical Name or Material Methyl acetate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.