missing translation for 'onlineSavingsMsg'
Learn More

Methyl Acetate, ReagentPlus™, 99%, Honeywell™

Product Code. 15661680 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
2.5 L
20 L
Packaging:
Glass Bottle
Stainless Steel Can
Unit Size:
2.5 L
20 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15661680 20 L Stainless Steel Can 20 L
15651680 2.5 L Glass Bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15661680 Supplier Honeywell Research Chemicals Supplier No. 18632520L
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Reagent Grade, 99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-20-9
Molecular Formula C3H6O2
Molecular Weight (g/mol) 74.079
MDL Number MFCD00008711
InChI Key KXKVLQRXCPHEJC-UHFFFAOYSA-N
Synonym tereton, devoton, acetic acid, methyl ester, methyl ethanoate, acetic acid methyl ester, methylacetat, acetate de methyle, methylacetaat, methyl acetic ester, octan metylu
PubChem CID 6584
ChEBI CHEBI:77700
IUPAC Name methyl acetate
SMILES CC(=O)OC

Specifications

Melting Point -98°C
Color Colorless
Density 0.934g/mL at 25°C
Boiling Point 58°C
Flash Point −13°C (8.6°F)
Packaging Stainless Steel Can
Linear Formula CH3COOCH3
Refractive Index n20/D 1.361(lit.)
Quantity 20 L
UN Number UN1231
Beilstein 1736662
Vapor Density 2.55 (vs air)
Formula Weight 74.08g/mol
Percent Purity 99%
Grade ReagentPlus
Physical Form Liquid
Chemical Name or Material Methyl Acetate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.