missing translation for 'onlineSavingsMsg'
Learn More

Methyl Acetate, TRC

Product Code. 30473223
Change view
Click to view available options
Quantity:
5 mL
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30473223 5 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30473223 Supplier Toronto Research Chemicals Supplier No. TRCM2753505ML

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methyl Acetate
CAS 79-20-9
Molecular Formula C3 H6 O2
Quantity 5 mL
Synonym Devoton, Methyl acetate, Methyl ethanoate, NSC 405071, Tereton
InChI Formula InChI=1S/C3H6O2/c1-3(4)5-2/h1-2H3
SMILES COC(=O)C
IUPAC Name methyl acetate
Molecular Weight (g/mol) 74.08
Formula Weight 74.0368
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.