missing translation for 'onlineSavingsMsg'
Learn More

Methyl acetoacetate, 99+%

Product Code. 10235270 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
Unit Size:
1 L
10 L
100mL
2.5 L
This item is not returnable. View return policy

Product Code. 10235270

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 105-45-3 | C5H8O3 | 116.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105-45-3
Molecular Formula C5H8O3
Molecular Weight (g/mol) 116.12
MDL Number MFCD00008784
InChI Key WRQNANDWMGAFTP-UHFFFAOYSA-N
Synonym methyl acetoacetate, acetoacetic acid methyl ester, methylacetoacetate, methyl 3-oxobutyrate, butanoic acid, 3-oxo-, methyl ester, methyl acetylacetate, methyl acetylacetonate, 3-oxobutanoic acid methyl ester, acetoacetic methyl ester, acetoacetic acid, methyl ester
PubChem CID 7757
IUPAC Name methyl 3-oxobutanoate
SMILES COC(=O)CC(C)=O

Specifications

Melting Point -28.0°C
Color Colorless
Density 1.0770g/mL
Boiling Point 169.0°C to 170.0°C
Flash Point 67°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula CH3COCH2CO2CH3
Refractive Index 1.4175 to 1.4295
Quantity 2.5 L
Beilstein 03, 632
Fieser 13,60
Merck Index 15, 6083
Specific Gravity 1.077
Solubility Information Solubility in water: 460g/L (20°C). Other solubilities: miscible with alcohol and ether,soluble in carbon tetrachloride
Viscosity 1.8 mPa.s (20°C)
Formula Weight 116.12
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material Methyl acetoacetate
Show More Show Less
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.