missing translation for 'onlineSavingsMsg'
Learn More

Methyl Acrylate (Stabilized), TRC

Product Code. 30135400
Change view
Click to view available options
Quantity:
25 mL
100 mL
250 mL
Unit Size:
100mL
250mL
25mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135400 250 mL 250mL
30134157 25 mL 25mL
30010772 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135400 Supplier Toronto Research Chemicals Supplier No. TRCM285060250ML
 Restricted Item
Estimated Shipment: 16-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96-33-3
Molecular Formula C4 H6 O2
Molecular Weight (g/mol) 86.09
Synonym Acrylic acid methyl ester (6CI,8CI), 2-Propenoic acid methyl ester, K 400, Methoxycarbonylethylene, Methyl acrylate, Methyl acrylic ester, Methyl prop-2-enoate, Methyl propenoate, NSC 24146
IUPAC Name methyl prop-2-enoate
SMILES COC(=O)C=C

Specifications

Quantity 250 mL
InChI Formula InChI=1S/C4H6O2/c1-3-4(5)6-2/h3H,1H2,2H3
Formula Weight 86.0368
Chemical Name or Material Acrylic acid methyl ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.