missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Adipoyl Chloride, TRC
Specifications
Specifications
| Chemical Name or Material | Methyl Adipoyl Chloride |
| CAS | 35444-44-1 |
| Molecular Formula | C7H11ClO3 |
| Quantity | 1 g |
| Synonym | 5-(Methoxycarbonyl)pentanoyl chloride, 5-Carbomethoxypentanoyl chloride, 5-Carbomethoxyvaleryl chloride, 5-Chlorocarbonylpentanoic acid methyl ester, 6-Chloro-6-oxohexanoic acid methyl ester, Adipic acid chloride methyl ester, Adipic acid methyl ester chloride, Adipic acid monomethyl ester chloride, Adipoyl chloride monomethyl ester, Methyl 5- (chlorocarbonyl)pentanoate, Methyl 5-(chloroformyl)pentanoate, Methyl 6-chloro-6-oxohexanoate, Methyl adipoyl chloride, Monomethyl adipoyl chloride |
| InChI Formula | InChI=1S/C7H11ClO3/c1-11-7(10)5-3-2-4-6(8)9/h2-5H2,1H3 |
| SMILES | ClC(CCCCC(OC)=O)=O |
| IUPAC Name | methyl 6-chloro-6-oxohexanoate |
| Molecular Weight (g/mol) | 178.61 |
| Formula Weight | 178.04 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?