missing translation for 'onlineSavingsMsg'
Learn More

Methyl Adipoyl Chloride, TRC

Product Code. 30473827
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Product Code. Quantity unitSize
30473827 500 mg 500mg
1 options
This item is not returnable. View return policy

Product Code. 30473827

Brand: Toronto Research Chemicals TRCM276200500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methyl Adipoyl Chloride
CAS 35444-44-1
Molecular Formula C7H11ClO3
Quantity 500 mg
Synonym 5-(Methoxycarbonyl)pentanoyl chloride, 5-Carbomethoxypentanoyl chloride, 5-Carbomethoxyvaleryl chloride, 5-Chlorocarbonylpentanoic acid methyl ester, 6-Chloro-6-oxohexanoic acid methyl ester, Adipic acid chloride methyl ester, Adipic acid methyl ester chloride, Adipic acid monomethyl ester chloride, Adipoyl chloride monomethyl ester, Methyl 5- (chlorocarbonyl)pentanoate, Methyl 5-(chloroformyl)pentanoate, Methyl 6-chloro-6-oxohexanoate, Methyl adipoyl chloride, Monomethyl adipoyl chloride
InChI Formula InChI=1S/C7H11ClO3/c1-11-7(10)5-3-2-4-6(8)9/h2-5H2,1H3
SMILES ClC(CCCCC(OC)=O)=O
IUPAC Name methyl 6-chloro-6-oxohexanoate
Molecular Weight (g/mol) 178.61
Formula Weight 178.04
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.