missing translation for 'onlineSavingsMsg'
Learn More

Methyl alpha-D-glucopyranoside, 98%

Product Code. 11454104
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11454104 100 g 100g
11464104 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11454104 Supplier Thermo Scientific Alfa Aesar Supplier No. A12484.22
 Restricted Item
Estimated Shipment: 07-08-2026
Aggiungi al carrello
Aggiungi al carrello
Request Bulk or Custom Format
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

CAS: 97-30-3 | C7H14O6 | 194.183 g/mol

Methyl alpha-D-glucopyranoside is a biochemical used to vary echo decay times in a study that assessed the effects of cryoprotection on the structure and activity of p21ras. It is also used in a study to investigate saccharide-mediated protection of chaotropic-induced deactivation of concanavalin A.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl alpha-D-glucopyranoside is a biochemical used to vary echo decay times in a study that assessed the effects of cryoprotection on the structure and activity of p21ras. It is also used in a study to investigate saccharide-mediated protection of chaotropic-induced deactivation of concanavalin A.

Solubility
Soluble in water. Insoluble in ether

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is hygroscopic in nature. Store under dry inert gas.
TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 97-30-3
Molecular Formula C7H14O6
Molecular Weight (g/mol) 194.183
MDL Number MFCD00064086
InChI Key HOVAGTYPODGVJG-IECVIRLLSA-N
Synonym methyl alpha-d-glucopyranoside, methyl alpha-d-glucoside, alpha-methylglucoside, alpha-methyl-d-glucoside, alpha-methyl d-glucose ether, methyl-alpha-d-glucopyranoside, alpha-d-methylglucoside, me alpha-glc, alpha-d-methyl glucoside, unii-qcf122nf3r
PubChem CID 7568545
IUPAC Name (2R,3R,4R,5R,6R)-2-(hydroxymethyl)-6-methoxyoxane-3,4,5-triol
SMILES COC1C(C(C(C(O1)CO)O)O)O

Specifica

Melting Point 166°C to 171°C
Density 1.46
Quantity 100 g
Beilstein 81568
Sensitivity Hygroscopic
Merck Index 14,6080
Solubility Information Soluble in water; Insoluble in ether
Optical Rotation +159° (c=10 in Water)
Formula Weight 194.18
Percent Purity ≥98%
Chemical Name or Material Methyl alpha-D-glucopyranoside
Vedi altri risultati Mostra meno risultati

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.