missing translation for 'onlineSavingsMsg'
Learn More

Methyl anthranilate, 99%

Product Code. 11457627
Change view
Click to view available options
Quantity:
250 g
500 g
1000 g
Unit Size:
1000g
250g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11457627 1000 g 1000g
11467627 250 g 250g
11438810 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11457627 Supplier Thermo Scientific Alfa Aesar Supplier No. A15681.0B
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 134-20-3 | C8H9NO2 | 151.17 g/mol

It is applied as a flavoring agent. Food companies use artificial MA in their candy and chewing gum to give them their grape flavor. Beverage companies have been adding it to the carbonated water and sugar in their soft drinks for the same reason.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 134-20-3
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.17
InChI Key VAMXMNNIEUEQDV-UHFFFAOYSA-N
IUPAC Name methyl 2-aminobenzoate
SMILES COC(=O)C1=CC=CC=C1N

Specifications

Melting Point 24°C
Boiling Point 260°C
Quantity 1000 g
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Methyl anthranilate
compliance-icons
Product Identifier
  • Methyl anthranilate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Methyl anthranilate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.