missing translation for 'onlineSavingsMsg'
Learn More

Methyl anthranilate, 99%

Product Code. 10580034 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 g
250 g
500 g
2.5 kg
Packaging:
Glass bottle
Unit Size:
2.5kg
250g
500g
50g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10580034 2.5 kg Glass bottle 2.5kg
10784071 50 g Glass bottle 50g
10469240 250 g Glass bottle 250g
10182980 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10580034 Supplier Thermo Scientific Acros Supplier No. 126320025
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 134-20-3 | C8H9NO2 | 151.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 134-20-3
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.17
MDL Number MFCD00007710
InChI Key VAMXMNNIEUEQDV-UHFFFAOYSA-N
Synonym methyl anthranilate, 2-aminobenzoic acid methyl ester, anthranilic acid methyl ester, methyl o-aminobenzoate, nevoli oil, methylanthranilate, o-carbomethoxyaniline, 2-carbomethoxyaniline, 2-methoxycarbonyl aniline, benzoic acid, 2-amino-, methyl ester
PubChem CID 8635
ChEBI CHEBI:73244
IUPAC Name methyl 2-aminobenzoate
SMILES COC(=O)C1=CC=CC=C1N

Specifications

Color Yellow to Brown
Density 1.1680g/mL
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula 2-(H2N)C6H4CO2CH3
Refractive Index 1.5810 to 1.5840 (20°C, 589nm)
Quantity 2.5 kg
Beilstein 14, 317
Merck Index 15, 6094
Formula Weight 151.16
Percent Purity ≥98.5%
Physical Form Liquid
Chemical Name or Material Methyl anthranilate, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.