missing translation for 'onlineSavingsMsg'
Learn More

Methyl Arachidate, 99%

Product Code. 15289836 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15289836 5 g 5g
15210846 1 g 1g
15299836 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15289836 Supplier Thermo Scientific Acros Supplier No. 452820050
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1120-28-1 | C21H42O2 | 326.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1120-28-1
Molecular Formula C21H42O2
Molecular Weight (g/mol) 326.57
MDL Number MFCD00009014
InChI Key QGBRLVONZXHAKJ-UHFFFAOYSA-N
Synonym methyl arachidate, eicosanoic acid, methyl ester, arachidic acid methyl ester, methyl eicosanoate, methyl arachisate, arachidic acid methylester, eicosanoic acid methyl ester, methylarachidate, methyl aracidate
PubChem CID 14259
IUPAC Name methyl icosanoate
SMILES CCCCCCCCCCCCCCCCCCCC(=O)OC

Specifications

Melting Point 45.0°C to 48.0°C
Boiling Point 215.0°C to 216.0°C (10.0 mmHg)
Flash Point >110°C
Assay Percent Range 98.5% min. (GC)
Linear Formula CH3(CH2)18COOCH3
Quantity 5 g
Formula Weight 326.56
Percent Purity 99%
Chemical Name or Material Methyl Arachidate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.