missing translation for 'onlineSavingsMsg'
Learn More

Methyl azetidine-3-carboxylate hydrochloride, 95%, Thermo Scientific Chemicals

Product Code. 15453456
Change view
Click to view available options
Quantity:
250 mg
500 mg
Unit Size:
250mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15453456 500 mg 500mg
15443456 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15453456 Supplier Thermo Scientific Alfa Aesar Supplier No. H57489.ME
 Restricted Item
Estimated Shipment: 23-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100202-39-9
Molecular Formula C5H10ClNO2
Molecular Weight (g/mol) 151.59
MDL Number MFCD01861758
InChI Key UOCWTLBPYROHEF-UHFFFAOYSA-N
Synonym methyl azetidine-3-carboxylate hydrochloride, azetidine-3-methyl carboxylate hydrochloride, azetidine-3-carboxylic acid methyl ester hydrochloride, 3-azetidinecarboxylic acid, methyl ester, hydrochloride, methyl azetidine-3-carboxylate hcl, methylazetidine-3-carboxylate hydrochloride, methyl 3-azetidinecarboxylate hydrochloride, azetidine-3-carboxylic acid methyl ester hcl, pubchem23516, acmc-209tka
PubChem CID 21100040
IUPAC Name methyl azetidine-3-carboxylate;hydrochloride
SMILES Cl.COC(=O)C1CNC1

Specifications

Boiling Point 187°C to 192°C
Flash Point 69°C (156°F)
Quantity 500 mg
Formula Weight 151.59
Percent Purity 95%
Chemical Name or Material Methyl azetidine-3-carboxylate hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.