missing translation for 'onlineSavingsMsg'
Learn More

Methyl Aziridine-2-carboxylate, TRC

Product Code. 30475871
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30475871 500 mg 500mg
30474717 50 mg 50mg
30475767 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30475871 Supplier Toronto Research Chemicals Supplier No. TRCM222485500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5950-34-5
Molecular Formula C4H7NO2
Molecular Weight (g/mol) 101.1
Synonym 2-Carbomethoxyaziridine
IUPAC Name methyl aziridine-2-carboxylate
SMILES COC(=O)C1CN1

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C4H7NO2/c1-7-4(6)3-2-5-3/h3,5H,2H2,1H3
Formula Weight 101.0477
Chemical Name or Material Methyl Aziridine-2-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.