missing translation for 'onlineSavingsMsg'
Learn More

Methyl behenate, 99%, analytical standard for GC

Product Code. p-7077211 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15592283

Brand: Thermo Scientific Acros 226810010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 929-77-1 | C23H46O2 | 354.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 929-77-1
Molecular Formula C23H46O2
Molecular Weight (g/mol) 354.62
MDL Number MFCD00009347
InChI Key QSQLTHHMFHEFIY-UHFFFAOYSA-N
Synonym methyl behenate, docosanoic acid, methyl ester, behenic acid methyl ester, docosanoic acid methyl ester, behenic acid, methyl ester, unii-04kbo9r771, behenic methyl ester, kemester 9022, docosanoic acid methyl, acmc-209rjf
PubChem CID 13584
IUPAC Name methyl docosanoate
SMILES CCCCCCCCCCCCCCCCCCCCCC(=O)OC

Specifications

Melting Point 54.0°C to 56.0°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass Bottle
Linear Formula CH3(CH2)20CO2CH3
Quantity 1 g
Merck Index 15,1022
Solubility Information (5% in Hexane) Clear colorless solution
Formula Weight 354.61
Percent Purity 99%
Chemical Name or Material Methyl behenate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.