missing translation for 'onlineSavingsMsg'
Learn More

Methyl Benzethonium chloride, MP Biomedicals™

Product Code. 17891381 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 g
25 g
Unit Size:
10g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17891381 25 g 25g
17881381 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17891381 Supplier MP Biomedicals Supplier No. 0219024025
  • ONLINE EXCLUSIVE PRICE
    1015.00 €
    959.70 € / 25g
    Save 55.30 €
    5% Off
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 25155-18-4
Molecular Formula C28H44ClNO2
Molecular Weight (g/mol) 462.115
InChI Key QWZLBLDNRUUYQI-UHFFFAOYSA-M
PubChem CID 5702238
IUPAC Name benzyl-dimethyl-[2-[2-[2-methyl-4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethoxy]ethyl]azanium;chloride
SMILES CC1=C(C=CC(=C1)C(C)(C)CC(C)(C)C)OCCOCC[N+](C)(C)CC2=CC=CC=C2.[Cl-]

Specifications

Color White
Quantity 25 g
Synonym methylbenzethonium chloride, methyl benzethonium chloride, unii-nn7590iuqx, nn7590iuqx, benzyldimethyl 2-2-2-methyl-4-2,4,4-trimethylpentan-2-yl phenoxy ethoxy ethyl azanium chloride, quat20, quat 20, prestwick_9, metilbenzetonio cloruro, benzyl-dimethyl-2-2-2-methyl-4-2,4,4-trimethylpentan-2-yl phenoxy ethoxy ethyl azanium;chloride
Formula Weight 462.1
Physical Form Powder
Chemical Name or Material Methyl Benzethonium chloride
compliance-icons compliance-icons
Product Identifier
  • Methyl Benzethonium Chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see on this label).
  • P330-Rinse mouth.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: methylbenzethonium chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.