missing translation for 'onlineSavingsMsg'
Learn More

Methyl benzo[b]thiophene-2-carboxylate, 97%, BLD Pharm™

Product Code. 30784084
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30784084 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30784084 Supplier BLD Pharm Supplier No. BD13996250mg

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 1444-65-1
Molecular Formula C12H14O
MDL Number MFCD00001629
Quantity 250 mg
SMILES O=C1CCCCC1C1=CC=CC=C1
Molecular Weight (g/mol) 174.24
Percent Purity 98%
Chemical Name or Material 2-Phenylcyclohexanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.