missing translation for 'onlineSavingsMsg'
Learn More

Methyl Benzoate, Extra Pure, SLR, Fisher Chemical™

Product Code. 10478600 Shop All Fisher Chemical Products
Click to view available options
Quantity:
500 mL
2.5 L
Unit Size:
2.5 L
500mL
This item is not returnable. View return policy

Product Code. 10478600

Brand: Fisher Chemical M/4460/PB17

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 93-58-3
Molecular Formula C8H8O2
Molecular Weight (g/mol) 136.15
MDL Number 8421
InChI Key QPJVMBTYPHYUOC-UHFFFAOYSA-N
Synonym methylbenzoate, benzoic acid, methyl ester, clorius, benzoic acid methyl ester, niobe oil, oil of niobe, methyl benzenecarboxylate, essence of niobe, oniobe oil, oxidate le
PubChem CID 7150
ChEBI CHEBI:72775
IUPAC Name methyl benzoate
SMILES COC(=O)C1=CC=CC=C1

Specifications

Melting Point -12°C
Color Yellow
Boiling Point 198°C
Packaging Plastic Coated Glass bottle
Quantity 2.5 L
Formula Weight 136.15g/mol
Physical Form Liquid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.