missing translation for 'onlineSavingsMsg'
Learn More

Methyl Benzoylformate, TRC

Product Code. 30464817
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30464817 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30464817 Supplier Toronto Research Chemicals Supplier No. TRCM3123401G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methyl 2-oxo-2-Phenylacetate (Methyl Benzoylformate)
CAS 15206-55-0
Molecular Formula C9 H8 O3
Quantity 1 g
Synonym Methyl 2-Oxo-2-phenylacetate, Benzeneacetic acid','α-oxo-','methyl ester, Glyoxylic acid','phenyl-','methyl ester (6CI,7CI,8CI), 2-Oxo-2-phenylacetic acid methyl ester, Benzoylformic acid methyl ester, Darocur MB, Darocur MBF, Doublecure 200, Genocure MBF, IHT-PI MBF, Irgacure MBF, JRCure 1055, MBF, MBF 1055, Methoxycarbonyl phenyl ketone, Methyl 2-oxo-2-phenylacetate, Methyl 2-phenyl-2-oxoacetate, Methyl benzoylformate, Methyl oxophenylacetate, Methyl phenyl-α-oxoacetate, Methyl phenylglyoxalate, Methyl phenylglyoxylate, Methyl phenyloxoacetate, Methyl α-oxobenzeneacetate, NSC 171206, NSC 409881, PI 55, Phenylglyoxylic acid methyl ester, Photoinitiator MBF, SB-PI 716, Speedcure MBF, Vicure 55, α-Oxobenzeneacetic acid methyl ester
InChI Formula InChI=1S/C9H8O3/c1-12-9(11)8(10)7-5-3-2-4-6-7/h2-6H,1H3
SMILES COC(=O)C(=O)c1ccccc1
IUPAC Name methyl 2-oxo-2-phenylacetate
Molecular Weight (g/mol) 164.16
Formula Weight 164.0473
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.