missing translation for 'onlineSavingsMsg'
Learn More

Methyl-beta-cyclodextrin, average Mw 1310

Product Code. 10673035 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10673035 5 g Glass bottle 5g
10717475 25 g Glass bottle 25g
10565734 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10673035 Supplier Thermo Scientific Acros Supplier No. 377110050
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 128446-36-6 | C54H94O35 | 1303.311 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chelation/Complexation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 128446-36-6
Molecular Formula C54H94O35
Molecular Weight (g/mol) 1303.311
MDL Number MFCD00074980
InChI Key YZOUYRAONFXZSI-SBHWVFSVSA-N
Synonym beta-cyclodextrin methyl ethers, beta-cyd, methyl-?cyclodextrin, beta-w7 m1.8, dimethyl-, a-cyclodextrin, .beta.-cyclodextrin,methylethers, methyl-b-cyclodextrin cell culture*tested
PubChem CID 51051622
SMILES COC1C(C2C(OC1OC3C(OC(C(C3OC)OC)OC4C(OC(C(C4OC)OC)OC5C(OC(C(C5OC)OC)OC6C(OC(C(C6OC)OC)OC7C(OC(C(C7OC)OC)OC8C(OC(O2)C(C8O)OC)CO)CO)CO)CO)CO)CO)CO)O

Specifications

Melting Point 165.0°C to 172.0°C
Color White
Density 187g/mL
Flash Point 187°C
Loss on Drying 5.0% max.
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 5 g
Specific Rotation +161° to +167° (20°C, 589nm) (C=5, water)
Solubility Information Solubility in water: 800g/L (25°C). Other solubilities: soluble in methanol,ethanol,acetone,pyridine,,dimethylsulfoxide and dimethylformamide
Physical Form Powder
Chemical Name or Material Methyl-β-cyclodextrin, Average Mn approx. 1310
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.