missing translation for 'onlineSavingsMsg'
Learn More

Methyl beta-D-galactopyranoside

Product Code. 11367995
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11367995 5 g 5g
11357995 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11367995 Supplier Thermo Scientific Alfa Aesar Supplier No. L08127.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1824-94-8 | C7H14O6 | 194.18 g/mol

It is a weak substrate and effective inducer of beta-D-galactosidase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a weak substrate and effective inducer of beta-D-galactosidase.

Solubility
Soluble in water and methanol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from heat. Keep it refrigerated.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1824-94-8
Molecular Formula C7H14O6
Molecular Weight (g/mol) 194.18
MDL Number MFCD00064357
InChI Key HOVAGTYPODGVJG-VOQCIKJUSA-N
Synonym methyl beta-d-galactopyranoside, methyl beta-d-galactoside, methyl galactoside, methylgalactoside, methyl beta-galactoside, galbetaome, unii-64ryd088rj, beta-d-galactopyranose methyl glycoside, beta-methyl-d-galactoside, methyl-beta-galactose
PubChem CID 94214
ChEBI CHEBI:17540
IUPAC Name (2R,3R,4S,5R,6R)-2-(hydroxymethyl)-6-methoxyoxane-3,4,5-triol
SMILES CO[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O

Specifications

Melting Point 175°C to 179°C
Quantity 5 g
Beilstein 81569
Solubility Information Soluble in water and methanol.
Optical Rotation −16° (c=1.5 in Methanol)
Formula Weight 194.18
Chemical Name or Material Methyl beta-D-galactopyranoside

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.