missing translation for 'onlineSavingsMsg'
Learn More

Methyl Beta-D-Galactopyranoside, TRC

Product Code. 30482826
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30482826 1 g 1g
30470970 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30482826 Supplier Toronto Research Chemicals Supplier No. TRCM3065001G
 Restricted Item
Estimated Shipment: 08-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1824-94-8
Molecular Formula C7 H14 O6
Molecular Weight (g/mol) 194.18
Synonym β-D-Galactopyranoside, methyl, Galactopyranoside, methyl, β-D- (8CI), Methyl β-D-galactopyranoside, 1-O-Methyl-β-D-galactopyranoside, Methyl galactoside, Methyl β-D-galactoside, Methyl β-galactoside, NSC 33685, β-Methylgalactose
IUPAC Name (2R,3R,4S,5R,6R)-2-(hydroxymethyl)-6-methoxyoxane-3,4,5-triol
SMILES CO[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C7H14O6/c1-12-7-6(11)5(10)4(9)3(2-8)13-7/h3-11H,2H2,1H3/t3-,4+,5+,6-,7-/m1/s1
Formula Weight 194.079
Chemical Name or Material Methyl beta-D-Galactopyranoside
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.