missing translation for 'onlineSavingsMsg'
Learn More

Methyl beta-D-ribofuranoside, 98%

Product Code. 15438349
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 15438349

Brand: Thermo Scientific Alfa Aesar J66828.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7473-45-2 | C6H12O5 | 164.157 g/mol

D-Ribose was converted into methyl 5-O-benzyl-beta-D-ribofuranoside and this, on tin-mediated allylation, gave a mixture of the 2-O-allyl and 3-O-allyl derivatives which were separated by chromatography. 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Ribose was converted into methyl 5-O-benzyl-beta-D-ribofuranoside and this, on tin-mediated allylation, gave a mixture of the 2-O-allyl and 3-O-allyl derivatives which were separated by chromatography. 

Solubility
Soluble in water

Notes
Store at 2-8°C. Incompatible with heat, oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7473-45-2
Molecular Formula C6H12O5
Molecular Weight (g/mol) 164.157
MDL Number MFCD00047075
InChI Key NALRCAPFICWVAQ-KVTDHHQDSA-N
Synonym methyl beta-d-ribofuranoside, methyl b-d-ribofuranoside, methyl, a-d-ribofuranoside, 2r,3s,4r,5r-2-hydroxymethyl-5-methoxytetrahydrofuran-3,4-diol, 2r,3s,4r,5r-2-hydroxymethyl-5-methoxyoxolane-3,4-diol, methyl pentofuranoside #, methyl .beta.-d-ribofuranoside, 1-o-methyl-beta-d-ribofuranoside, pubchem10614, methyl-b-d-ribofuranoside
PubChem CID 81983
IUPAC Name (2R,3S,4R,5R)-2-(hydroxymethyl)-5-methoxyoxolane-3,4-diol
SMILES COC1C(C(C(O1)CO)O)O

Specifications

Melting Point 78°C
Color White
Quantity 25 g
Solubility Information Soluble in water
Optical Rotation −50° (c=2 in Water)
Formula Weight 164.16
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Methyl beta-D-ribofuranoside

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.