missing translation for 'onlineSavingsMsg'
Learn More

Methyl bromoacetate, 99%

Product Code. 10072230 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Unit Size:
100g
2.5kg
25g
500g
This item is not returnable. View return policy

Product Code. 10072230

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 96-32-2 | C3H5BrO2 | 152.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96-32-2
Molecular Formula C3H5BrO2
Molecular Weight (g/mol) 152.98
MDL Number MFCD00000189
InChI Key YDCHPLOFQATIDS-UHFFFAOYSA-N
Synonym methyl bromoacetate, bromoacetic acid methyl ester, methyl monobromoacetate, acetic acid, bromo-, methyl ester, methyl alpha-bromoacetate, methylbromoacetate, acetic acid, 2-bromo-, methyl ester, methylester kyseliny bromoctove, methyl .alpha.-bromoacetate, methylester kyseliny bromoctove czech
PubChem CID 60984
IUPAC Name methyl 2-bromoacetate
SMILES COC(=O)CBr

Specifications

Color Colorless to Yellow
Density 1.6100g/mL
Boiling Point 145.0°C
Flash Point 62°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula BrCH2CO2CH3
Refractive Index 1.4570 to 1.4590
Quantity 25 g
Beilstein 02, 213
Specific Gravity 1.61
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in methanol, ether and acetone
Formula Weight 152.98
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Methyl bromoacetate, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Methyl bromoacetate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 C
  • Skin sensitiser Category 1
Hazard Statement
  • H301-Toxic if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Methyl bromoacetate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.