missing translation for 'onlineSavingsMsg'
Learn More

Methyl alpha-bromophenylacetate, 97+%

Product Code. 10346294 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
25 g
Unit Size:
1g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10346294 25 g 25g
10623975 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10346294 Supplier Thermo Scientific Acros Supplier No. 379300250
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 3042-81-7 | C9H9BrO2 | 229.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3042-81-7
Molecular Formula C9H9BrO2
Molecular Weight (g/mol) 229.07
MDL Number MFCD00013535
InChI Key NHFBYYMNJUMVOT-UHFFFAOYSA-N
Synonym methyl alpha-bromophenylacetate, methyl bromophenylacetate, methyl bromo phenyl acetate, methyl .alpha.-bromophenylacetate, methyl2-bromo-2-phenylacetate, methyl .alpha.-bromobenzeneacetate, benzeneacetic acid, .alpha.-bromo-, methyl ester, +/--methyl alpha-bromophenylacetate, alpha-bromophenylacetic acid methyl ester, methyl .alpha.-bromo-.alpha.-phenylacetate
PubChem CID 137806
IUPAC Name methyl 2-bromo-2-phenylacetate
SMILES COC(=O)C(C1=CC=CC=C1)Br

Specifications

Density 1.4550g/mL
Boiling Point 113.0°C (3.0 mmHg)
Flash Point 113°C
Assay Percent Range 97% min. (GC)
Packaging Glass bottle
Linear Formula C6H5CH(Br)CO2CH3
Quantity 25 g
Specific Gravity 1.455
Solubility Information Solubility in water: .. Other solubilities: soluble in most organic solvents
Formula Weight 229.07
Percent Purity 97+%
Chemical Name or Material Methyl α-bromophenylacetate
Show More Show Less
compliance-icons
Product Identifier
  • Methyl alpha-bromophenylacetate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.