missing translation for 'onlineSavingsMsg'
Learn More

Methyl butyrate, 98+%

Product Code. 11386856
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11386856 500 mL 500mL
11376856 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11386856 Supplier Thermo Scientific Alfa Aesar Supplier No. L04250.AP
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 623-42-7 | C5H10O2 | 102.13 g/mol

Methyl butyrate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl butyrate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 623-42-7
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.13
MDL Number MFCD00009391
InChI Key UUIQMZJEGPQKFD-UHFFFAOYSA-N
Synonym methyl butyrate, butanoic acid, methyl ester, methyl n-butyrate, butyric acid, methyl ester, methyl n-butanoate, butyric acid methyl ester, methyl-n-butyrate, n-butyric acid methyl ester, methyl butyrate natural, unii-cgx598508o
PubChem CID 12180
IUPAC Name methyl butanoate
SMILES CCCC(=O)OC

Specifications

Melting Point -85°C to -84°C
Density 0.898
Boiling Point 100°C to 102°C
Flash Point 11°C (51°F)
Refractive Index 1.386
Quantity 500 mL
UN Number UN1237
Beilstein 1740743
Merck Index 14,6035
Solubility Information Slightly soluble in water.
Formula Weight 102.14
Percent Purity ≥98%
Chemical Name or Material Methyl butyrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.