missing translation for 'onlineSavingsMsg'
Learn More

Methyl butyrylacetate, 98+%

Product Code. 11352277
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11352277 25 g 25g
11362277 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11352277 Supplier Thermo Scientific Alfa Aesar Supplier No. L09826.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 30414-54-1 | C7H12O3 | 144.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30414-54-1
Molecular Formula C7H12O3
Molecular Weight (g/mol) 144.17
MDL Number MFCD00040995
InChI Key SJPCQNABHNCLPB-UHFFFAOYSA-N
Synonym methyl butyrylacetate, methyl 3-oxocaproate, 3-oxohexanoic acid methyl ester, hexanoic acid, 3-oxo-, methyl ester, methyl 3-ketohexanoate, butyrylacetic acid methyl ester, 3-oxo-hexanoic acid methyl ester, bem, acmc-1ahmy, ksc497e5t
PubChem CID 121700
IUPAC Name methyl 3-oxohexanoate
SMILES CCCC(=O)CC(=O)OC

Specifications

Melting Point -43°C
Density 1.037
Boiling Point 85°C to 90°C (5 mmHg)
Flash Point 90°C (194°F)
Refractive Index 1.426
Quantity 25 g
Beilstein 636165
Formula Weight 144.17
Percent Purity ≥98%
Chemical Name or Material Methyl butyrylacetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.