missing translation for 'onlineSavingsMsg'
Learn More

Methyl carbamate, 99%

Product Code. 11447847
Click to view available options
Quantity:
50 g
100 g
500 g
Unit Size:
100g
500g
50g
This item is not returnable. View return policy

Product Code. 11447847

Brand: Thermo Scientific Alfa Aesar A16523.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 598-55-0 | C2H5NO2 | 75.07 g/mol

Methyl carbamate was used in the synthesis of protected aminocyclopropanes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl carbamate was used in the synthesis of protected aminocyclopropanes.

Solubility
Soluble in water(20g/L).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, metals, alkali metals.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 598-55-0
Molecular Formula C2H5NO2
Molecular Weight (g/mol) 75.07
MDL Number MFCD00007964
InChI Key GTCAXTIRRLKXRU-UHFFFAOYSA-N
Synonym urethylane, carbamic acid, methyl ester, methylurethan, carbamic acid methyl ester, methylkarbamat, methylkarbamat czech, unii-9wfx634x2t, ccris 885, methylester kyseliny karbaminove, methylester kyseliny karbaminove czech
PubChem CID 11722
ChEBI CHEBI:76606
IUPAC Name methyl carbamate
SMILES COC(N)=O

Specifications

Melting Point 54°C to 56°C
Boiling Point 176°C to 178°C
Beilstein 635779
Merck Index 14,6036
Solubility Information Soluble in water(20g/L).
Density 1.14
Quantity 100 g
Flash Point >100°C (212°F)
Formula Weight 75.07
Percent Purity 99%
Chemical Name or Material Methyl carbamate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.