missing translation for 'onlineSavingsMsg'
Learn More

Methyl cellulose, MP Biomedicals™

Product Code. 11487792 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 g
1 kg
Unit Size:
1kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11487792 1 kg 1kg
11477792 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11487792 Supplier MP Biomedicals Supplier No. 0215549691
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Methyl cellulose , White Powder

  • It is used as a substitute for water-soluble gums,to render paper grease proof in adhesives,as thickening agent in cosmetics,as protective colloid in emulsions,as binder and stabilizer in foods,as bulk producer in the formulation of dietetic foods,Pharmaceutic aid (suspending agent),Laxative.
  • It is a chemical compound derived from cellulose. It is a hydrophilic white powder in pure form and dissolves in cold (but not in hot) water, forming a clear viscous solution or gel.
  • It is used as a thickener and emulsifier in various food and cosmetic products. It is not digestible, not toxic, and not an allergen.

Chemical Identifiers

CAS 9004-67-5
Molecular Formula C20H38O11
MDL Number MFCD00081763
Synonym Cellulose methyl ether, Methocel MC, Cellothyl, Syncelose

Specifications

Color White
pH 6.7
Loss on Drying ≤5.0%
Quantity 1 kg
Solubility Information Soluble in cold water. Insoluble in hot water.
InChI Key YLGXILFCIXHCMC-UHFFFAOYNA-N
Molecular Weight (g/mol) 454.51
Physical Form Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.