missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Methyl cellulose, viscosity 400 cPs

Product Code. 11344828
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11344828 2.5 kg 2.5kg
11324828 100 g 100g
11334828 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11344828 Supplier Thermo Scientific Chemicals Supplier No. 043147.A4
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 9004-67-5 | C20H38O11 | 454.513 g/mol

Methyl cellulose is used as a thickener and emulsifier in various food and cosmetic products like hair shampoos, toothpastes and liquid soaps. It is used in the preparation of capsules in nutritional supplements. It is also used as a gel in advanced cookery and as a lubricant. It is also involved in the treatment of constipation. It acts as a buffer additive in capillary electrophoresis to control electoosmotic flow for improved separations. Further, it is used as a performance additive in construction materials and as sizing in the production of papers and textiles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9004-67-5
Molecular Formula C20H38O11
Molecular Weight (g/mol) 454.513
MDL Number MFCD00081763
InChI Key YLGXILFCIXHCMC-JHGZEJCSSA-N
PubChem CID 51063134
IUPAC Name (5R)-2,3,4-trimethoxy-6-(methoxymethyl)-5-[(2S)-3,4,5-trimethoxy-6-(methoxymethyl)oxan-2-yl]oxyoxane
SMILES COCC1C(C(C(C(O1)OC2C(OC(C(C2OC)OC)OC)COC)OC)OC)OC

Specifications

Odor Odorless
Quantity 2.5 kg
Merck Index 14,6040
Solubility Information Soluble in cold water.
Physical Form Powder
Chemical Name or Material Methyl cellulose, Viscosity: 400cPs

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.