missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Methyl cellulose, viscosity 4000 cP (2% solution in water)

Product Code. 10522174 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Plastic drum
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10522174 2.5 kg Plastic drum 2.5kg
10287200 100 g Plastic bottle 100g
10429940 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10522174 Supplier Thermo Scientific Chemicals Supplier No. 258110025
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 9004-67-5 | C20H38O11 | 454.513 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9004-67-5
Molecular Formula C20H38O11
Molecular Weight (g/mol) 454.513
MDL Number MFCD00081763
InChI Key YLGXILFCIXHCMC-JHGZEJCSSA-N
PubChem CID 51063134
IUPAC Name (5R)-2,3,4-trimethoxy-6-(methoxymethyl)-5-[(2S)-3,4,5-trimethoxy-6-(methoxymethyl)oxan-2-yl]oxyoxane
SMILES COCC1C(C(C(C(O1)OC2C(OC(C(C2OC)OC)OC)COC)OC)OC)OC

Specifications

Color Gray-White
Physical Form Powder
Merck Index 15, 6111
Quantity 2.5 kg
Infrared Spectrum Authentic
Loss on Drying 5% max.
Packaging Plastic drum
Viscosity 3000 to 5600cP (2% solution in water,20°C)
Chemical Name or Material Methyl cellulose, Viscosity (2% Solution in Water) 3000-5600 CP

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.