Learn More
Thermo Scientific Chemicals Methyl cellulose, viscosity 8000 cPs
Description
Methyl cellulose is used as a thickener and emulsifier in various food and cosmetic products, and also as a treatment of constipation. It is also used as binders, lubricants, steric stabilizer, water retention aid and sizing in the production of papers and textiles as it protects the fibers from absorbing water or oil.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Odor | Odorless |
| Quantity | 100 g |
| Merck Index | 14,6040 |
| Solubility Information | Soluble in glacial acetic acid and cold water. Insoluble in hot water,ethanol,ether and chloroform. |
| Physical Form | Powder |
| Chemical Name or Material | Methyl cellulose, Viscosity: 8000cPs |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.