missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Methyl cellulose, viscosity 8000 cPs

Product Code. 11335807
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11335807 500 g 500g
11315807 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11335807 Supplier Thermo Scientific Chemicals Supplier No. 043483.36
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 9004-67-5 | C20H38O11 | 454.513 g/mol

Methyl cellulose is used as a thickener and emulsifier in various food and cosmetic products, and also as a treatment of constipation. It is also used as binders, lubricants, steric stabilizer, water retention aid and sizing in the production of papers and textiles as it protects the fibers from absorbing water or oil.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 9004-67-5
Molecular Formula C20H38O11
Molecular Weight (g/mol) 454.513
MDL Number MFCD00081763
InChI Key YLGXILFCIXHCMC-JHGZEJCSSA-N
PubChem CID 51063134
IUPAC Name (5R)-2,3,4-trimethoxy-6-(methoxymethyl)-5-[(2S)-3,4,5-trimethoxy-6-(methoxymethyl)oxan-2-yl]oxyoxane
SMILES COCC1C(C(C(C(O1)OC2C(OC(C(C2OC)OC)OC)COC)OC)OC)OC

Specifikationer

Odor Odorless
Quantity 500 g
Merck Index 14,6040
Solubility Information Soluble in glacial acetic acid and cold water. Insoluble in hot water,ethanol,ether and chloroform.
Physical Form Powder
Chemical Name or Material Methyl cellulose, Viscosity: 8000cPs

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.