missing translation for 'onlineSavingsMsg'
Learn More

Methyl Chloroacetate, MP Biomedicals

Product Code. 11453615 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11453615 1 kg 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11453615 Supplier MP Biomedicals Supplier No. 0521688925
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

CAS 96-34-4
Molecular Formula C3H5ClO2
Quantity 1 kg
Synonym methyl chloroacetate, chloroacetic acid methyl ester, methyl monochloroacetate, methyl chloroethanoate, acetic acid, chloro-, methyl ester, methyl monochloracetate, methyl alpha-chloroacetate, methylchloroacetate, unii-450vsb182i, ccris 7749
InChI Key QABLOFMHHSOFRJ-UHFFFAOYSA-N
SMILES COC(=O)CCl
IUPAC Name methyl 2-chloroacetate
Molecular Weight (g/mol) 108.521
PubChem CID 7295
Formula Weight 108.53
Chemical Name or Material Methyl Chloroacetate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.