missing translation for 'onlineSavingsMsg'
Learn More

Methyl cholate, 97%

Product Code. 11321837
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11321837 25 g 25g
11311837 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11321837 Supplier Thermo Scientific Alfa Aesar Supplier No. L05129.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1448-36-8 | C25H42O5 | 422.606 g/mol

Methyl cholate, is used as a bile acid derivative as anti-inflammatory and antitumor agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl cholate, is used as a bile acid derivative as anti-inflammatory and antitumor agent.

Solubility
Insoluble in water. Soluble in medium polarity organic solvents.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1448-36-8
Molecular Formula C25H42O5
Molecular Weight (g/mol) 422.606
MDL Number MFCD00064934
InChI Key DLYVTEULDNMQAR-BSEHQKOQSA-N
Synonym Cholic acid methyl ester
PubChem CID 133124937
IUPAC Name methyl (4S)-4-[(3R,7S,8S,9R,10R,12R,13S,14R,17S)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoate
SMILES CC(CCC(=O)OC)C1CCC2C1(C(CC3C2C(CC4C3(CCC(C4)O)C)O)O)C

Specifications

Melting Point 155°C to 156°C
Quantity 25 g
Beilstein 2226989
Merck Index 14,2203
Solubility Information Insoluble in water. Soluble in medium polarity organic solvents.
Formula Weight 422.61
Percent Purity ≥98%
Chemical Name or Material Methyl cholate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.