missing translation for 'onlineSavingsMsg'
Learn More

Methyl cis-11-eicosenoate, 99%

Product Code. 10893951 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10893951 1 g Glass bottle 1g
10894141 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10893951 Supplier Thermo Scientific Acros Supplier No. 428530010
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2390-09-2 | C21H40O2 | 324.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2390-09-2
Molecular Formula C21H40O2
Molecular Weight (g/mol) 324.55
MDL Number MFCD00010458
InChI Key RBKMRGOHCLRTLZ-ZHACJKMWSA-N
Synonym cis-11-eicosenoic acid methyl ester, cis-methyl 11-eicosenoate, 11-eicosenoic acid, methyl ester, z, methyl cis-11-eicosenoate, methyl 11z-icos-11-enoate, methyl z-icos-11-enoate, z-methyl icos-11-enoate, cis-11-eicosenoic acid, methyl ester, 11z-11-icosenoic acid methyl ester
PubChem CID 5463047
IUPAC Name methyl (Z)-icos-11-enoate
SMILES CCCCCCCCC=CCCCCCCCCCC(=O)OC

Specifications

Melting Point -15.0°C
Flash Point >113°C
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)7CH=CH(CH2)9CO2CH3
Quantity 1 g
Formula Weight 324.55
Percent Purity 99%
Chemical Name or Material Methyl cis-11-eicosenoate
compliance-icons
Product Identifier
  • Methyl cis-11-eicosenoate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.