missing translation for 'onlineSavingsMsg'
Learn More

Methyl-β-cyclodextrin, MedChemExpress

Product Code. 16490759
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
50 g
Unit Size:
10g
1g
25g
50g
5g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16490759 1 g 1g
16453378 10 g 10g
16439906 50 g 50g
16416107 5 g 5g
16402746 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16490759 Supplier Medchem Express Supplier No. HY101461
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Methyl-β-cyclodextrin (Methyl-beta-cyclodextrin) is a cyclic heptasaccharide used to deliver hydrophobic drugs based on its property of solubilizing non-polar substances. Methyl-β-cyclodextrin is also extensively used as a cholesterol-depleting reagent. Methyl-β-cyclodextrin strongly reduces clathrin-dependent endocytosis.

Methyl-β-cyclodextrin (Methyl-beta-cyclodextrin) is a cyclic heptasaccharide used to deliver hydrophobic drugs based on its property of solubilizing non-polar substances. Methyl-β-cyclodextrin is also extensively used as a cholesterol-depleting reagent. Methyl-β-cyclodextrin strongly reduces clathrin-dependent endocytosis.

Specifications

Chemical Name or Material Methyl-β-cyclodextrin
CAS 128446-36-6
Color White
Purity Grade Notes Research
Quantity 1 g
Synonym Methyl-beta-cyclodextrin
Solubility Information DMSO : ≥ 100 mg/mL ∣H2O : ≥ 50 mg/mL
SMILES CO[C@@H]1[C@H](O[R])[C@H](O[R])[C@@H](C)C(CO[R])O1.[7].[R=H or -CH3]
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.