missing translation for 'onlineSavingsMsg'
Learn More

Methyl cyclohex-1-enecarboxylate, 98%, BLD Pharm™

Product Code. 30804064
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
100 g
Unit Size:
100g
10g
1g
25g
5g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30804064 1 g 1g
30796953 10 g 10g
30801619 25 g 25g
30796179 5 g 5g
30793229 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 30804064 Supplier BLD Pharm Supplier No. BD1379331g
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Especificaciones

CAS 18448-47-0
Molecular Formula C8H12O2
MDL Number MFCD00001544
Quantity 1 g
SMILES COC(=O)C1=CCCCC1
Molecular Weight (g/mol) 140.18
Percent Purity 98%
Chemical Name or Material Methyl cyclohex-1-enecarboxylate
Título del producto
Seleccione un problema

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.