missing translation for 'onlineSavingsMsg'
Learn More

Methyl Cyclopropane Carboxylate, TRC

Product Code. 30482450
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
Product Code. Quantity unitSize
30482450 50 g 50g
1 options
This item is not returnable. View return policy

Product Code. 30482450

Brand: Toronto Research Chemicals TRCM29471050G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Cyclopropanecarboxylic Acid Methyl Ester
CAS 2868-37-3
Molecular Formula C5 H8 O2
Quantity 50 g
Synonym Cyclopropanecarboxylic acid, methyl ester, Cyclopropanecarboxylic acid methyl ester, Cyclopropanoic acid methyl ester, Methyl cyclopropanecarboxylate, Methyl cyclopropylcarboxylate
InChI Formula InChI=1S/C5H8O2/c1-7-5(6)4-2-3-4/h4H,2-3H2,1H3
SMILES COC(=O)C1CC1
IUPAC Name methyl cyclopropanecarboxylate
Molecular Weight (g/mol) 100.116
Formula Weight 100.052
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.