missing translation for 'onlineSavingsMsg'
Learn More

Methyl α-D-Mannopyranoside, 99.3%, MP Biomedicals

Product Code. 11417382 Shop All MP Biomedicals Products
Click to view available options
Quantity:
10 g
25 g
100 g
Unit Size:
100g
10g
25g

Product Code. 11417382

Brand: MP Biomedicals 0215393710

Please to purchase this item. Need a web account? Register with us today!

Chemical Identifiers

CAS 617-04-9
Molecular Formula C7H14O6
Molecular Weight (g/mol) 194.18
MDL Number MFCD00063262
InChI Key HOVAGTYPODGVJG-UHFFFAOYNA-N
Synonym methyl a-d-mannopyranoside, methyl alpha-d-mannopyranoside, methyl alpha-d-mannoside, alpha-methyl-d-mannoside, alpha-methyl mannopyranoside, 1-o-methyl-alpha-d-mannopyranoside, methyl-alpha-d-mannoside, alpha-d-methyl mannoside, o1-methyl-mannose, alpha-methyl-d---mannoside
PubChem CID 101798
ChEBI CHEBI:43943
IUPAC Name 2-(hydroxymethyl)-6-methoxyoxane-3,4,5-triol
SMILES COC1OC(CO)C(O)C(O)C1O

Specifications

Color White
Assay Percent Range ∽ 99%
Quantity 10 g
Formula Weight 194.2
Physical Form Crystalline Powder
Chemical Name or Material Methyl α-D-Mannopyranoside
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.