missing translation for 'onlineSavingsMsg'
Learn More

Methyl α-D-mannopyranoside, ≥99%, Fisher BioReagents™

Product Code. 11413084
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
500 g
Packaging:
Amber Glass
Unit Size:
100g
10g
25g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11413084 500 g Amber Glass 500g
10596184 10 g Amber Glass 10g
10286094 25 g Amber Glass 25g
11403084 100 g Amber Glass 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11413084 Supplier Fisher BioReagents Supplier No. BP2530500
  • ONLINE EXCLUSIVE PRICE
    995.00 €
    941.00 € / 500g
    Save 54.00 €
    5% Off
 Restricted Item
Estimated Shipment: 11-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 617-04-9 | C7H14O6 | 194.18 g/mol

Methyl α-D-mannopyranoside, ≥99%

  • Methyl α-D-mannopyranoside is a lectin inhibitory sugar
  • Competitive inhibitor of the binding of mannose by E. coli
  • Intended for research use only
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 617-04-9
Molecular Formula C7H14O6
Molecular Weight (g/mol) 194.18
MDL Number MFCD00063262
InChI Key HOVAGTYPODGVJG-UHFFFAOYNA-N
Synonym methyl a-d-mannopyranoside, methyl alpha-d-mannopyranoside, methyl alpha-d-mannoside, alpha-methyl-d-mannoside, alpha-methyl mannopyranoside, 1-o-methyl-alpha-d-mannopyranoside, methyl-alpha-d-mannoside, alpha-d-methyl mannoside, o1-methyl-mannose, alpha-methyl-d---mannoside
PubChem CID 101798
ChEBI CHEBI:43943
SMILES COC1OC(CO)C(O)C(O)C1O

Specifications

Melting Point 193°C to 196°C
Color White
Loss on Drying < 0.5%
Assay Percent Range ≥99%
Packaging Amber Glass
Quantity 500 g
Specific Rotation 76 to 85° (+ or -)
IUPAC Name (2R,3S,4S,5S,6S)-2-(hydroxymethyl)-6-methoxyoxane-3,4,5-triol
Formula Weight 194.2
Percent Purity ≥99%
Physical Form Powder or Solid
Chemical Name or Material Methyl α-D-Mannopyranoside
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.