missing translation for 'onlineSavingsMsg'
Learn More

alpha-Methyl-D-mannopyranoside, 99+%

Product Code. 10720575 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10720575 500 g Glass bottle 500g
10229873 25 g Glass bottle 25g
10023170 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10720575 Supplier Thermo Scientific Acros Supplier No. 229255000
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

alpha-Methyl-D-mannopyranoside, 99+%, Quantity: 500g, Packaging: Glass bottle, Gray or White, Melting Point: 186.0 deg.C to 194.0 deg.C, Molecular Weight: 194.18, Percent Purity: 99+%, Appearance: (1% aq. soln.) clear, colorless, Assay Percent Range: 99% min. (ELSD) (HPLC) | CAS: 617-04-9 | C7H14O6 | 194.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 617-04-9
Molecular Formula C7H14O6
Molecular Weight (g/mol) 194.18
MDL Number MFCD00063262
InChI Key HOVAGTYPODGVJG-UHFFFAOYNA-N
Synonym methyl a-d-mannopyranoside, methyl alpha-d-mannopyranoside, methyl alpha-d-mannoside, alpha-methyl-d-mannoside, alpha-methyl mannopyranoside, 1-o-methyl-alpha-d-mannopyranoside, methyl-alpha-d-mannoside, alpha-d-methyl mannoside, o1-methyl-mannose, alpha-methyl-d---mannoside
PubChem CID 101798
ChEBI CHEBI:43943
IUPAC Name (2R,3S,4S,5S,6S)-2-(hydroxymethyl)-6-methoxyoxane-3,4,5-triol
SMILES COC1OC(CO)C(O)C(O)C1O

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 186.0°C to 194.0°C
Color Gray or White
Infrared Spectrum Authentic
Assay Percent Range 99% min. (ELSD) (HPLC)
Packaging Glass bottle
Quantity 500 g
Specific Rotation Condition + 79.00 (20.00°C c=10,H2O)
Specific Rotation + 79.00
Solubility Information (10% in water) Clear colorless
Formula Weight 194.18
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material α-Methyl-D-mannopyranoside
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.