missing translation for 'onlineSavingsMsg'
Learn More

Methyl α-D-Mannopyranoside, MP Biomedicals™

Product Code. 11447382 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11447382 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11447382 Supplier MP Biomedicals Supplier No. 0215393790
  • ONLINE EXCLUSIVE PRICE
    659.00 €
    624.75 € / 500g
    Save 34.25 €
    5% Off
 Restricted Item
Estimated Shipment: 05-03-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Reduces Con A binding to the lymphocyte surface in culture. Methyl α-D-Mannopyranoside, MP Biomedicals a competitor inhibitor of the binding of mannose by Escherichia coli; it was used in a study to investigate the primary mannose binding site of pr

Methyl α-D-mannopyranoside has been used to synthesize a series of tri- and tetrahydroxylated seven-membered iminosugars in a study that worked towards a stable noeuromycin analog with a D-manno configuration.

Specifications

CAS 617-04-9
Melting Point 187°C to 197°C
Molecular Formula C7H14O6
MDL Number MFCD00063262
Quantity 500 g
Synonym methyl a-d-mannopyranoside, methyl alpha-d-mannopyranoside, methyl alpha-d-mannoside, alpha-methyl-d-mannoside, alpha-methyl mannopyranoside, 1-o-methyl-alpha-d-mannopyranoside, methyl-alpha-d-mannoside, alpha-d-methyl mannoside, o1-methyl-mannose, alpha-methyl-d---mannoside
Solubility Information Soluble in water.
InChI Key HOVAGTYPODGVJG-UHFFFAOYNA-N
SMILES COC1OC(CO)C(O)C(O)C1O
IUPAC Name (2R,3S,4S,5S,6S)-2-(hydroxymethyl)-6-methoxyoxane-3,4,5-triol
Molecular Weight (g/mol) 194.18
PubChem CID 101798
ChEBI CHEBI:43943
Formula Weight 194.20
Percent Purity ∼99%
Grade Not Available
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.