missing translation for 'onlineSavingsMsg'
Learn More

Methyl decanoate, 98+%

Product Code. 11487577
Change view
Click to view available options
Quantity:
50 g
100 g
250 g
Unit Size:
100g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11487577 250 g 250g
11477577 50 g 50g
11428810 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11487577 Supplier Thermo Scientific Alfa Aesar Supplier No. A15658.30
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 110-42-9 | C11H22O2 | 186.30 g/mol

It is employed as an intermediate for caproic acid, detergents, emulsifiers, wetting agents, stabilizers, resins, lubricants, plasticizers and flavoring. As a chemical intermediate for alkanolamides, fatty alcohols & acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as an intermediate for caproic acid, detergents, emulsifiers, wetting agents, stabilizers, resins, lubricants, plasticizers and flavoring. As a chemical intermediate for alkanolamides, fatty alcohols & acids.

Solubility
Soluble in alcohol. Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-42-9
Molecular Formula C11H22O2
Molecular Weight (g/mol) 186.30
MDL Number MFCD00009580
InChI Key YRHYCMZPEVDGFQ-UHFFFAOYSA-N
Synonym methyl caprate, decanoic acid, methyl ester, methyl caprinate, capric acid methyl ester, decanoic acid methyl ester, methyl-n-caprate, uniphat a30, metholene 2095, methyl n-decanoate, methyl n-caprate
PubChem CID 8050
IUPAC Name methyl decanoate
SMILES CCCCCCCCCC(=O)OC

Specifications

Melting Point -18°C
Density 0.877
Boiling Point 223°C to 225°C
Flash Point 94°C (201°F)
Refractive Index 1.425
Quantity 250 g
Beilstein 1759170
Solubility Information Soluble in alcohol. Insoluble in water.
Formula Weight 186.3
Percent Purity 99%
Chemical Name or Material Methyl decanoate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.