missing translation for 'onlineSavingsMsg'
Learn More

Methyl Decanoate(Methyl Caprate), TRC

Product Code. 30009129
Change view
Click to view available options
Quantity:
2.5 g
25 g
Unit Size:
2.5g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30009129 2.5 g 2.5g
30005523 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30009129 Supplier Toronto Research Chemicals Supplier No. TRCM2948452.5G
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-42-9
Molecular Formula C11 H22 O2
Molecular Weight (g/mol) 186.29
Synonym C 1098, CE 1095, Capric acid methyl ester, Estasol MD 10, Metholene 2095, Methyl caprate, Methyl caprinate, Methyl decanoate, Methyl n-caprate, Methyl n-decanoate, NSC 3713, Pastell M 10, Uniphat A 30
IUPAC Name methyl decanoate
SMILES CCCCCCCCCC(=O)OC

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C11H22O2/c1-3-4-5-6-7-8-9-10-11(12)13-2/h3-10H2,1-2H3
Formula Weight 186.162
Chemical Name or Material Capric Acid Methyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.