missing translation for 'onlineSavingsMsg'
Learn More

Methyl dodecanoate, 99%

Product Code. p-7020509
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11424124

Brand: Thermo Scientific Alfa Aesar A12492.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 111-82-0 | C13H26O2 | 214.349 g/mol

Methyl dodecanoate used in production of biodiesel. It has been used in selective synthesis of the secondary amide surfactant, N-methyl lauroylethanolamide. It is also utilized as emollients, perfuming agents skin conditioning.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl dodecanoate used in production of biodiesel. It has been used in selective synthesis of the secondary amide surfactant, N-methyl lauroylethanolamide. It is also utilized as emollients, perfuming agents skin conditioning.

Solubility
Not miscible in water. Soluble in alcohol, dipropylene glycol.

Notes
Store at room temperature. Ensure good ventilation.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-82-0
Molecular Formula C13H26O2
Molecular Weight (g/mol) 214.349
MDL Number MFCD00008966
InChI Key UQDUPQYQJKYHQI-UHFFFAOYSA-N
Synonym methyl laurate, lauric acid methyl ester, dodecanoic acid, methyl ester, methyl laurinate, methyl dodecylate, methyl n-dodecanoate, uniphat a40, metholene 2296, lauric acid, methyl ester, stepan c40
PubChem CID 8139
ChEBI CHEBI:87494
IUPAC Name methyl dodecanoate
SMILES CCCCCCCCCCCC(=O)OC

Specifications

Melting Point 4°C to 5°C
Density 0.870
Boiling Point 261°C to 262°C
Flash Point 125°C (257°F)
Assay Percent Range 99%
Refractive Index 1.4325
Quantity 5 g
Beilstein 1767780
Solubility Information Not miscible in water. Soluble in alcohol,dipropylene glycol.
Formula Weight 214.35
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Methyl Laurate
Show More Show Less
compliance-icons
Product Identifier
  • Methyl dodecanoate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P391-Collect spillage.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Dodecanoic acid, methyl ester

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.