missing translation for 'onlineSavingsMsg'
Learn More

Methyl Gallate, TRC

Product Code. 17951615
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17951615 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17951615 Supplier Toronto Research Chemicals Supplier No. TRCM3115105G
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 99-24-1
Molecular Formula C8 H8 O5
Synonym Benzoic acid, 3,4,5-trihydroxy-, methyl ester, Gallic acid, methyl ester (6CI,8CI), 3,4,5-Trihydroxybenzoic acid methyl ester, Gallincin, Methyl 3,4,5-trihydroxybenzoate, Methyl gallate, NSC 363001
InChI Formula InChI=1S/C8H8O5/c1-13-8(12)4-2-5(9)7(11)6(10)3-4/h2-3,9-11H,1H3
SMILES COC(=O)c1cc(O)c(O)c(O)c1
IUPAC Name methyl 3,4,5-trihydroxybenzoate
Molecular Weight (g/mol) 184.15
Formula Weight 184.0372
Quantity 5 g
Chemical Name or Material Methylgallate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.